Learn More
Glass Expansion Quartz Injector 1.2mm
SDP
Catalog No. 501907220
Encompass_Preferred
Quartz Injector with 1.2mm ID.A 1.2mm ID Quartz Injector (48807022), designed for volatile organic solvents and non-HF matrices. Compatible with Spectro Blue SOP Semi Demountable Torch (SDT) 50-190-7218 (31-808-3405), Spectro Blue EOP SDT 50-190-7217 (31-808-3394) and Spectro Blue TI SDT 50-190-7225 (31-808-3468).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7220 Supplier Glass Expansion Supplier No. 318083410
Quartz Injector with 1.2mm ID.A 1.2mm ID Quartz Injector (48807022), designed for volatile organic solvents and non-HF matrices. Compatible with Spectro Blue SOP Semi Demountable Torch (SDT) 50-190-7218 (31-808-3405), Spectro Blue EOP SDT 50-190-7217 (31-808-3394) and Spectro Blue TI SDT 50-190-7225 (31-808-3468).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001