Learn More
Glass Expansion Quartz Injector 3.0mm SpectroBlue EOP & TI
SDP
Catalog No. 502703131
Encompass_Preferred
Quartz Injector with 3.0mm ID (48807025).A 3.0mm ID Quartz Injector, designed for high TDS samples and non-HF matrices. Compatible with Spectro Blue SOP Semi Demountable Torch (SDT) 50-190-7218 (31-808-3405), Spectro Blue EOP SDT 50-190-7217 (31-808-3394) and Spectro Blue TI SDT 50-190-7225 (31-808-3468).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3131 Supplier Glass Expansion Supplier No. 318083412
Quartz Injector with 3.0mm ID (48807025).A 3.0mm ID Quartz Injector, designed for high TDS samples and non-HF matrices. Compatible with Spectro Blue SOP Semi Demountable Torch (SDT) 50-190-7218 (31-808-3405), Spectro Blue EOP SDT 50-190-7217 (31-808-3394) and Spectro Blue TI SDT 50-190-7225 (31-808-3468).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001