Learn More
Perkin Elmer US LLC Quartz Injector for Demountable ESI ZipTorch, 1.5 mm I.D.
Catalog No. 502104525
Encompass_Preferred
This Demountable Injector is for use with Avio 200 and 500 ICP-OES series instruments. It is designed for use with the quartz, organics and silicon nitride demountable Elemental Scientific ZipTorches. The demountable design allows the injector to be removed and replaced efficiently for cleaning without dismounting the torch.
Catalog No. 50-210-4525 Supplier Perkin Elmer US LLC Supplier No. N0791860
This Demountable Injector is for use with Avio 200 and 500 ICP-OES series instruments. It is designed for use with the quartz, organics and silicon nitride demountable Elemental Scientific ZipTorches. The demountable design allows the injector to be removed and replaced efficiently for cleaning without dismounting the torch.