Learn More
Perkin Elmer US LLC Quartz Injector for Demountable ZipTorch, 2.0 mm I.D. Optima 8x00
Catalog No. 502113141
Encompass_Preferred
This Demountable Injector is for use with Optima 8x00 ICP-OES series instruments. It is designed for use with the quartz, organics and silicon nitride demountable ZipTorches. The demountable design allows the injector to be removed and replaced efficiently for cleaning without dismounting the torch.
Catalog No. 50-211-3141 Supplier Perkin Elmer US LLC Supplier No. N0782009
This Demountable Injector is for use with Optima 8x00 ICP-OES series instruments. It is designed for use with the quartz, organics and silicon nitride demountable ZipTorches. The demountable design allows the injector to be removed and replaced efficiently for cleaning without dismounting the torch.