Learn More
Glass Expansion Quartz Injector 0.8mm ID PerkinElmer Avio 200/500
SDP
Catalog No. 502703114
Encompass_Preferred
Quartz Tapered Injector with 0.8mm ID. 0.8mm ID tapered quartz injector (N0791185) suitable for high-volatility organics and non-HF matrices. For use with Avio 200/220 and 500/550 ICP-OES series. Quartz injectors are designed for organic solvents and non-HF matrices. Compatible with PerkinElmer Avio 200/220 D-Torch (P/N 50-190-7038), Avio 500/550 D-Torch (P/N 50-190-7039), Avio 200/220 Adjustable torch body (P/N 50-211-3618) and Avio 500/550 Adjustable torch body (P/N 50-211-2626).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3114 Supplier Glass Expansion Supplier No. 318083785
Quartz Tapered Injector with 0.8mm ID. 0.8mm ID tapered quartz injector (N0791185) suitable for high-volatility organics and non-HF matrices. For use with Avio 200/220 and 500/550 ICP-OES series. Quartz injectors are designed for organic solvents and non-HF matrices. Compatible with PerkinElmer Avio 200/220 D-Torch (P/N 50-190-7038), Avio 500/550 D-Torch (P/N 50-190-7039), Avio 200/220 Adjustable torch body (P/N 50-211-3618) and Avio 500/550 Adjustable torch body (P/N 50-211-2626).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001