Learn More
Glass Expansion Quartz Tapered Injector with 1.0mm ID. A 1.0mm ID quartz tapered injector, ideal for use with volatile organic solvent-based matrices. Compatible with PerkinElmer ICP-MS NexION 1000/2000/5000 D-Torch (30-808-3927).
SDP
Catalog No. 502703130
Encompass_Preferred
Quartz Tapered Injector with 1.0mm ID. A 1.0mm ID quartz tapered injector, ideal for use with volatile organic solvent-based matrices. Compatible with PerkinElmer ICP-MS NexION 1000/2000/5000 D-Torch (30-808-3927). Not suitable for use with HF. When replacing your injector, it is recommended to replace the injector ferrule (31-808-2962), on average every 6 months. Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3130 Supplier Glass Expansion Supplier No. 318083423
Quartz Tapered Injector with 1.0mm ID. A 1.0mm ID quartz tapered injector, ideal for use with volatile organic solvent-based matrices. Compatible with PerkinElmer ICP-MS NexION 1000/2000/5000 D-Torch (30-808-3927). Not suitable for use with HF. When replacing your injector, it is recommended to replace the injector ferrule (31-808-2962), on average every 6 months. Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001