Learn More
Glass Expansion Quartz Tapered Injector with 1.2mm ID. 1.2mm ID tapered quartz injector (N0775226). This quartz injector is for use with PerkinElmer Optima 2x00/4x00/5x00/7x00 DV ICP-OES series instruments.
SDP
Catalog No. 502703156
Encompass_Preferred
Quartz Tapered Injector with 1.2mm ID. 1.2mm ID tapered quartz injector (N0775226). This quartz injector is for use with PerkinElmer Optima 2x00/4x00/5x00/7x00 DV ICP-OES series instruments. They are designed for organic solvents and non-HF matrices. A variety of sizes are available as an alternative to the standard injector.PerkinElmer Optima 2x00/4x00/5x00/7x00 DV ICP-OES series Injectors - Compatible with 50-190-7006 (30-808-0845), 50-190-7010 (30-808-1122), 50-190-7019 (30-808-1470), 50-190-7022 (30-808-2926), 50-904-7597 (N0772005), 50-904-3780 (N0770338, PG31016), 50-904-3781 (N0770344).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3156 Supplier Glass Expansion Supplier No. 318081154
Quartz Tapered Injector with 1.2mm ID. 1.2mm ID tapered quartz injector (N0775226). This quartz injector is for use with PerkinElmer Optima 2x00/4x00/5x00/7x00 DV ICP-OES series instruments. They are designed for organic solvents and non-HF matrices. A variety of sizes are available as an alternative to the standard injector.PerkinElmer Optima 2x00/4x00/5x00/7x00 DV ICP-OES series Injectors - Compatible with 50-190-7006 (30-808-0845), 50-190-7010 (30-808-1122), 50-190-7019 (30-808-1470), 50-190-7022 (30-808-2926), 50-904-7597 (N0772005), 50-904-3780 (N0770338, PG31016), 50-904-3781 (N0770344).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001