Learn More
Glass Expansion Quartz Mini Torch, Shimadzu 9000/9800 (30-808-4220, 211-81448, 21181448)
SDP
Catalog No. 501906987
Encompass_Preferred
Quartz Mini Torch, single-piece, 1.5 mm injector (211-81448), standard for Shimadzu 9000/9800 Series ICP-OES.Suitable for routine aqueous analysis, but not inert to samples containing hydrofluoric acid (HF). For high-salt solution sample quartz torch with 1.8 mm see catalog number 50-190-6983. For high-salt solution sample quartz torch with 2.0 mm injector ID see catalog number 50-190-6998.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-6987 Supplier Glass Expansion Supplier No. 308070534
Quartz Mini Torch, single-piece, 1.5 mm injector (211-81448), standard for Shimadzu 9000/9800 Series ICP-OES.Suitable for routine aqueous analysis, but not inert to samples containing hydrofluoric acid (HF). For high-salt solution sample quartz torch with 1.8 mm see catalog number 50-190-6983. For high-salt solution sample quartz torch with 2.0 mm injector ID see catalog number 50-190-6998.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001