Learn More
Glass Expansion Quartz Outer Tube for D-Torch
SDP
Supplier:  Glass Expansion 318083062
Encompass_Preferred
Quartz Outer Tube for D-Torch. Compatible with Agilent (Varian) 700-ES Series Radial, Vista Radial ICP-OES D-Torch (30-808-3052, 501907026). The D-Torch, provides a robust & high-performing alternative to both single-piece and semi-demountable quartz torches, ensuring greater robustness & a lower cost of ownership. With the D-Torch design, the analyst most often replaces only the outer tube, rather than replacing the entire torch or a quartz torch body.When not in use, clean as per recommended guidelines, & store in original box.If analyzing high salt, fusions, wear metals in oil, hydrofluoric acid (HF) or volatile organics, we recommend the ceramic outer tube P/N 501907180, which does not devitrify or crack from thermal shock, like quartz. Compared to quartz outer tubes, the ceramic alternative has an extended lifespan, significantly reducing maintenance, cleaning, and downtime caused by torch failure.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7177