Learn More
Glass Expansion Quartz Outer Tube for D-Torch
Supplier: Glass Expansion 318082888
Quartz Outer Tube for D-Torch. Compatible with Horiba 24, 38 & 138 Series, Ultima, Activa ICP-OES D-Torch (30-808-2882). The D-Torch, provides a robust and high-performing alternative to both single-piece and semi-demountable quartz torches, ensuring greater robustness & a lower cost of ownership. With the D-Torch design, the analyst most often replaces only the outer tube, rather than replacing the entire torch or a quartz torch body.When not in use, clean as per recommended guidelines, and store in original box.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001