Learn More
Glass Expansion Quartz Inner Tube
SDP
Supplier:  Glass Expansion 318089200
Encompass_Preferred
Quartz Inner Tube. Compatible with Thermo (TJA) Iris, Atomscan 16 & 25, 61E, Intrepid Radial ICP-OES, Glass Expansion's Fully Demountable PT2000 Torch (30-808-0041), which is obsolete.The fully demountable torch, provides a robust and high-performing alternative to both single-piece and semi-demountable quartz torches, ensuring greater robustness and a lower cost of ownership. With the fully demountable torch design, the analyst most often replaces only the outer tube, rather than replacing the entire torch or a quartz torch body.When not in use, clean as per recommended guidelines, and store in original box.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7280