Learn More
Glass Expansion Quartz Outer Tube
SDP
Supplier:  Glass Expansion 318080707
Encompass_Preferred
Quartz Outer Tube. Compatible with Thermo (TJA) Iris, Tracescan, Trace, Advantage, Intrepid Axial ICP-OES, ABC Fully Demountable Torch (30-808-0704). The fully demountable torch, provides a robust and high-performing alternative to both single-piece and semi-demountable quartz torches, ensuring greater robustness and a lower cost of ownership. With the fully demountable torch design, the analyst most often replaces only the outer tube, rather than replacing the entire torch or a quartz torch body.When not in use, clean as per recommended guidelines, and store in original box.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7101