Learn More
Thermo Scientific Quartz Reactor 18 MM DIAM 1 EA
Supplier:  Thermo Scientific 46820070
Encompass
Cylindrical quartz vessel with inner diameter of 18 millimeters used as chemical reactor in laboratory settings for analytical scale chemistry experiments and procedures due to inertness to many chemicals, ability to withstand high temperatures, and ultraviolet light transmission; provides small reaction volume suitable for analytical procedures.
Catalog No. 50-134-7442