Learn More
Perkin Elmer US LLC Quartz Sample Tube
Catalog No. 502113437
Encompass_Preferred
The Quartz Sample Tube is used in the standard DMA 7/7e furnace, this tube is a component of the Quartz Parallel Plate Systems Kit (N5390135).
Catalog No. 50-211-3437 Supplier Perkin Elmer US LLC Supplier No. N5191837
The Quartz Sample Tube is used in the standard DMA 7/7e furnace, this tube is a component of the Quartz Parallel Plate Systems Kit (N5390135).