Learn More
Glass Expansion Quartz Spray Chamber with B14 drain for Shimadzu ICPM-2030
SDP
Catalog No. 501906969
Encompass_Preferred
Quartz Spray Chamber with B14 drain for Shimadzu ICPM-2030 (211-93577). Gravity drain quartz Twister spray chamber with B14 drain & Helix CT. Includes Helix CT locking screw & seal (1 pc), & JC-13 Metal Ball Joint Clip (1 pc). Made from quartz, 50mL volume. Compatible with Shimadzu ICPM-2030 ICP-MS.The Twister features a baffle, which efficiently removes larger droplets, producing a smaller mean droplet size entering the plasma. Resulting in a more robust plasma, making the Twister an excellent choice for the best possible precision & for both high TDS or organic sample matrices.The unique Helix CT interface consists of two parts, a ConstantTorque Locking Screw & chemically inert PressFit ferrule. The main feature of the Helix CT is the built-in, ratcheting torque control mechanism that allows for a consistent, zero dead volume, inert seal of the nebulizer.Replacement Helix CT Seals (PKT 4) NC2081825.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-6969 Supplier Glass Expansion Supplier No. 218093992
Quartz Spray Chamber with B14 drain for Shimadzu ICPM-2030 (211-93577). Gravity drain quartz Twister spray chamber with B14 drain & Helix CT. Includes Helix CT locking screw & seal (1 pc), & JC-13 Metal Ball Joint Clip (1 pc). Made from quartz, 50mL volume. Compatible with Shimadzu ICPM-2030 ICP-MS.The Twister features a baffle, which efficiently removes larger droplets, producing a smaller mean droplet size entering the plasma. Resulting in a more robust plasma, making the Twister an excellent choice for the best possible precision & for both high TDS or organic sample matrices.The unique Helix CT interface consists of two parts, a ConstantTorque Locking Screw & chemically inert PressFit ferrule. The main feature of the Helix CT is the built-in, ratcheting torque control mechanism that allows for a consistent, zero dead volume, inert seal of the nebulizer.Replacement Helix CT Seals (PKT 4) NC2081825.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001