Learn More
Perkin Elmer US LLC Quartz SSI Cyclonic/Scott Dual Spray Chamber with Gas Port for NexION 300/350 PC3 Systems
Catalog No. 502105174
Encompass_Preferred
The SSI Cyclonic/Scott Dual Spray Chamber, made of quartz, is designed to homogenize sample aerosol prior to introduction to the ICP-MS instrument. It features a secure, leak-free threaded drain port, O-ring-free connections to the nebulizer and torch injector, internal baffle, and threaded auxiliary port for gas addition.
Catalog No. 50-210-5174 Supplier Perkin Elmer US LLC Supplier No. N8145313
Add to Cart
Add to Cart
Request a Quote
The SSI Cyclonic/Scott Dual Spray Chamber, made of quartz, is designed to homogenize sample aerosol prior to introduction to the ICP-MS instrument. It features a secure, leak-free threaded drain port, O-ring-free connections to the nebulizer and torch injector, internal baffle, and threaded auxiliary port for gas addition.