Learn More
Perkin Elmer US LLC Quartz SUPRASIL Flowcell - 10 mm, 440 µL
Catalog No. 502112317
Encompass_Preferred
Fluorescence Spectroscopy Flow-Through cells are used for measuring samples with continuous flow or with sippers where individual samples are aspirated into the cell. All cells have a center height of 15 mm.
Catalog No. 50-211-2317 Supplier Perkin Elmer US LLC Supplier No. N4202039
Add to Cart
Add to Cart
Request a Quote
Fluorescence Spectroscopy Flow-Through cells are used for measuring samples with continuous flow or with sippers where individual samples are aspirated into the cell. All cells have a center height of 15 mm.