Learn More
Perkin Elmer US LLC Quartz Suprasil Macro Cuvette with PTFE Stopper - 10 mm, Qty. 2
Catalog No. 502093283
Encompass_Preferred
These durable, fire-fused cells are standard rectangular spectrophotometer cuvettes with two optically-polished windows. Their stoppers provide an air-tight seal. They are made from the highest quality quartz and glass material available to ensure excellent transmission values and highly accurate results. Features of PerkinElmer Cuvettes: Material Path Length Outer Length Outer Width Capacity Size UV Quartz 10 mm 12.5 mm 12.5 mm 3.5 mL Macro
Catalog No. 50-209-3283 Supplier Perkin Elmer US LLC Supplier No. N4103017
These durable, fire-fused cells are standard rectangular spectrophotometer cuvettes with two optically-polished windows. Their stoppers provide an air-tight seal. They are made from the highest quality quartz and glass material available to ensure excellent transmission values and highly accurate results. Features of PerkinElmer Cuvettes: Material Path Length Outer Length Outer Width Capacity Size UV Quartz 10 mm 12.5 mm 12.5 mm 3.5 mL Macro