Learn More
Glass Expansion Quartz Torch with 0.8mm Injector for Agilent (Varian) 700-ES or Vista Radial
SDP
Catalog No. 502703170
Encompass_Preferred
Quartz Torch, single-piece, 0.8 mm ID injector, for Agilent (Varian) 700-ES or Vista Radial Series ICP-OES. Recommended for routine volatile organic solvent analysis but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3170 Supplier Glass Expansion Supplier No. 308070539
Quartz Torch, single-piece, 0.8 mm ID injector, for Agilent (Varian) 700-ES or Vista Radial Series ICP-OES. Recommended for routine volatile organic solvent analysis but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001