Learn More
Glass Expansion Quartz Torch with 1.0mm Injector for Agilent7500
SDP
Catalog No. 501906982
Encompass_Preferred
Quartz Torch, single-piece, 1.0 mm id injector, for petrochemical and volatile organic solvent analysis with Agilent 7500 ICP-MS. Suitable for organic applications with volatile solvents but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-6982 Supplier Glass Expansion Supplier No. 308070516
Quartz Torch, single-piece, 1.0 mm id injector, for petrochemical and volatile organic solvent analysis with Agilent 7500 ICP-MS. Suitable for organic applications with volatile solvents but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001