Learn More
Glass Expansion Quartz Torch-Low Flow with 1.4mm Injector for 700ES or Vista Radial
SDP
Catalog No. 501906975
Encompass_Preferred
Quartz Torch (Low Flow), single-piece, with 1.4 mm ID injector, for Agilent (Varian) 700-ES or Vista Radial Series ICP-OES (2010069600). Recommended for less-volatile organic solvent analysis but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-6975 Supplier Glass Expansion Supplier No. 308000015
Quartz Torch (Low Flow), single-piece, with 1.4 mm ID injector, for Agilent (Varian) 700-ES or Vista Radial Series ICP-OES (2010069600). Recommended for less-volatile organic solvent analysis but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001