Learn More
Glass Expansion Quartz Torch, High TDS, 1.8mm , Shimadzu 7500/8100/9000/9800
SDP
Catalog No. 501906983
Encompass_Preferred
Quartz Torch, single-piece, 1.8 mm injector (211-74323), standard for Shimadzu 7500/8100/9000/9800 Series ICP-OES.Suitable for high-salt solution sample analysis, but not inert to samples containing hydrofluoric acid (HF). For high-salt solution sample quartz torch with 2.0 mm injector ID see P/N 501906998.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-6983 Supplier Glass Expansion Supplier No. 308070518
Quartz Torch, single-piece, 1.8 mm injector (211-74323), standard for Shimadzu 7500/8100/9000/9800 Series ICP-OES.Suitable for high-salt solution sample analysis, but not inert to samples containing hydrofluoric acid (HF). For high-salt solution sample quartz torch with 2.0 mm injector ID see P/N 501906998.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001