Learn More
Glass Expansion Quartz Torch with 2.0mm Injector for X Series
SDP
Catalog No. 501906986
Encompass_Preferred
Quartz Torch, single-piece, 2.0 mm id injector, for routine aqueous analysis with Thermo X-Series ICP-MS (3601147 and 3600791). Suitable for routine aqueous applications but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-6986 Supplier Glass Expansion Supplier No. 308070532
Quartz Torch, single-piece, 2.0 mm id injector, for routine aqueous analysis with Thermo X-Series ICP-MS (3601147 and 3600791). Suitable for routine aqueous applications but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001