Learn More
Glass Expansion Quartz Torch 2.3mm Injector for 700-ES or Vista Axial
SDP
Catalog No. 501906974
Encompass_Preferred
Quartz Torch, single-piece, 2.3 mm ID injector , for Agilent (Varian) 700-ES, Liberty or Vista Series ICP-OES (2010081200). Suitable for routine aqueous analysis but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-6974 Supplier Glass Expansion Supplier No. 308000014
Quartz Torch, single-piece, 2.3 mm ID injector , for Agilent (Varian) 700-ES, Liberty or Vista Series ICP-OES (2010081200). Suitable for routine aqueous analysis but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001