Learn More
Glass Expansion Quartz Torch 2.5mm injector with gas fittings
SDP
Catalog No. 501906984
Encompass_Preferred
Quartz Torch, single-piece, 2.5 mm ID injector with gas fittings, for Spectro (Ametek) SOP Modula, CIROS, Genesis, ARCOS Series ICP-OES (S75008). Suitable for routine aqueous analysis but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-6984 Supplier Glass Expansion Supplier No. 308070524
Quartz Torch, single-piece, 2.5 mm ID injector with gas fittings, for Spectro (Ametek) SOP Modula, CIROS, Genesis, ARCOS Series ICP-OES (S75008). Suitable for routine aqueous analysis but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001