Learn More
Glass Expansion Quartz Torch High Solids with Sheath Gas for 700-ES or Vista Axial
SDP
Catalog No. 501906985
Encompass_Preferred
Quartz Torch, single-piece (High Solids with Sheath Gas), 2.4 mm ID injector (2010122400). Recommended for high total dissolved solids (TDS) analysis with Agilent (Varian) axial 700-ES, Liberty and Vista Series ICP-OES. Used with auxillary gas module (AGM) 1 for control of sheath gas flow, provides better performance than the standard high solids torch. Suitable for routine analysis of high TDS up to 25% but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-6985 Supplier Glass Expansion Supplier No. 308070526
Quartz Torch, single-piece (High Solids with Sheath Gas), 2.4 mm ID injector (2010122400). Recommended for high total dissolved solids (TDS) analysis with Agilent (Varian) axial 700-ES, Liberty and Vista Series ICP-OES. Used with auxillary gas module (AGM) 1 for control of sheath gas flow, provides better performance than the standard high solids torch. Suitable for routine analysis of high TDS up to 25% but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001