Learn More
Glass Expansion Quartz Torch, High Solids, with Ball Joint and 1.8mm Injector for 700-ES or Vista Radial
SDP
Catalog No. 501906988
Encompass_Preferred
Quartz Torch (High Solids), single-piece, with Ball Joint and 1.8 mm ID injector, for Agilent (Varian) 700-ES or Vista Radial Series ICP-OES (CP914870). Recommended for aqueous samples and less-volatile organic solvent analysis but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-6988 Supplier Glass Expansion Supplier No. 308070536
Quartz Torch (High Solids), single-piece, with Ball Joint and 1.8 mm ID injector, for Agilent (Varian) 700-ES or Vista Radial Series ICP-OES (CP914870). Recommended for aqueous samples and less-volatile organic solvent analysis but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001