Learn More
Glass Expansion Quartz Torch with 3.0mm Injector, Sheath Gas & Fittings for Spectro EOP, SOP
SDP
Catalog No. 5000556649
Encompass_Preferred
Quartz Torch with 3.0mm Injector, Sheath Gas & Fittings for Spectro EOP, SOP Sourced from Glass Expansion
Catalog No. 50-005-56649 Supplier Glass Expansion Supplier No. 308070570
Quartz Torch with 3.0mm Injector, Sheath Gas & Fittings for Spectro EOP, SOP Sourced from Glass Expansion