Learn More
Glass Expansion Quartz Torch with 3 slots for Optima 4300/5300/7300 V
SDP
Catalog No. 501906981
Encompass_Preferred
Quartz Torch, 3-Slot demountable quartz torch standard for PerkinElmer Optima 4300/5300/7300 V Series ICP-OES (N0771500).Recommended for organic solvent applications, but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-6981 Supplier Glass Expansion Supplier No. 308070064
Quartz Torch, 3-Slot demountable quartz torch standard for PerkinElmer Optima 4300/5300/7300 V Series ICP-OES (N0771500).Recommended for organic solvent applications, but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001