Learn More
Glass Expansion Quartz Torch with 1.5mm Injector for Bruker/Varian ICP-MS
SDP
Catalog No. 501906997
Encompass_Preferred
Quartz Torch, single-piece, 1.5 mm id injector, for aqueous and less-volatile organic solvent analysis with Bruker/Varian ICP-MS (2010123600). Suitable for routine aqueous and less-volatile organic solvent applications but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-6997 Supplier Glass Expansion Supplier No. 308070562
Quartz Torch, single-piece, 1.5 mm id injector, for aqueous and less-volatile organic solvent analysis with Bruker/Varian ICP-MS (2010123600). Suitable for routine aqueous and less-volatile organic solvent applications but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001