Learn More
Glass Expansion Quartz Torch 2.5mm injector, 4mm OD side arms for Spectro EOP
SDP
Catalog No. 501907003
Encompass_Preferred
Quartz Torch, single-piece, 2.5 mm injector, 4mm OD side arms for Spectro EOP ICP-OES (48105052).Suitable for routine aqueous analysis, but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7003 Supplier Glass Expansion Supplier No. 308080317
Quartz Torch, single-piece, 2.5 mm injector, 4mm OD side arms for Spectro EOP ICP-OES (48105052).Suitable for routine aqueous analysis, but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001