Learn More
Glass Expansion Quartz Torch with 2.5mm Injector for Agilent 7500, 2 Projections, HMI Compatible
SDP
Catalog No. 501906990
Encompass_Preferred
Quartz Torch, single-piece, 2.5 mm id injector, 2 Projections, HMI Compatible and for most routine sample analysis with Agilent 7500 ICP-MS. Standard quartz torch suitable for animal feed, chemicals and fertilizers, clinical and forensic materials, drinking, ground and surface water, food and drink, geological without HF, metals, plants, soil and sediment without HF, wastewater and sludge, brines and salts, isotopic analysis of minerals, but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-6990 Supplier Glass Expansion Supplier No. 308070542
Quartz Torch, single-piece, 2.5 mm id injector, 2 Projections, HMI Compatible and for most routine sample analysis with Agilent 7500 ICP-MS. Standard quartz torch suitable for animal feed, chemicals and fertilizers, clinical and forensic materials, drinking, ground and surface water, food and drink, geological without HF, metals, plants, soil and sediment without HF, wastewater and sludge, brines and salts, isotopic analysis of minerals, but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001