Learn More
Glass Expansion Quartz Torch for Nu Plasma with 1.5mm Injector
SDP
Catalog No. 501906996
Encompass_Preferred
Quartz Torch, single-piece, 1.5 mm injector (319-308), standard for Nu Plasma HR & Nu Plasma 3 ICP-MS.Suitable for routine aqueous analysis and organic applications, but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-6996 Supplier Glass Expansion Supplier No. 308070560
Quartz Torch, single-piece, 1.5 mm injector (319-308), standard for Nu Plasma HR & Nu Plasma 3 ICP-MS.Suitable for routine aqueous analysis and organic applications, but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001