Learn More
Glass Expansion Quartz Mini Torch for Shimadzu ICPMS-2030 (211-94446, 211-94018)
SDP
Catalog No. 501906999
Encompass_Preferred
Quartz Mini Torch, single-piece, 1.1 mm injector (211-94446, 211-94018), standard for Shimadzu 2030 ICP-MS.Suitable for routine aqueous analysis, but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-6999 Supplier Glass Expansion Supplier No. 308070567
Quartz Mini Torch, single-piece, 1.1 mm injector (211-94446, 211-94018), standard for Shimadzu 2030 ICP-MS.Suitable for routine aqueous analysis, but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001