Learn More
Glass Expansion Quartz Torch with 3 slots for Optima 4300/5300/7300 V (long life)
SDP
Catalog No. 501907033
Encompass_Preferred
Quartz Torch (long life), 3-Slot demountable quartz torch standard for PerkinElmer Optima 4300/5300/7300 V Series ICP-OES (N0771561).Recommended for organic solvent applications, but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7033 Supplier Glass Expansion Supplier No. 308083593
Quartz Torch (long life), 3-Slot demountable quartz torch standard for PerkinElmer Optima 4300/5300/7300 V Series ICP-OES (N0771561).Recommended for organic solvent applications, but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001