Learn More
Glass Expansion Quartz Torch with 2.5mm Injector for Agilent7500
SDP
Catalog No. 501907005
Encompass_Preferred
Quartz Torch, single-piece, 2.5 mm id injector, for Agilent 7500 ICP-MS. Suitable for aqueous samples with high total dissolved solids, but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7005 Supplier Glass Expansion Supplier No. 308080816
Quartz Torch, single-piece, 2.5 mm id injector, for Agilent 7500 ICP-MS. Suitable for aqueous samples with high total dissolved solids, but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001