Learn More
Glass Expansion Quartz Torch - High Solids for 700-ES or Vista Radial
SDP
Catalog No. 501907011
Encompass_Preferred
Quartz Torch, single-piece, 1.8 mm ID injector (2010095000), for high total dissolved solids (TDS) analysis with Agilent (Varian) 700-ES, Liberty or Vista Series ICP-OES. Suitable for routine analysis of high TDS up to 25% but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7011 Supplier Glass Expansion Supplier No. 308081182
Quartz Torch, single-piece, 1.8 mm ID injector (2010095000), for high total dissolved solids (TDS) analysis with Agilent (Varian) 700-ES, Liberty or Vista Series ICP-OES. Suitable for routine analysis of high TDS up to 25% but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001