Learn More
Glass Expansion Quartz Torch 2.5mm injector with gas fittings for Spectro EOP
SDP
Catalog No. 501907014
Encompass_Preferred
Quartz Torch, single-piece, 2.5 mm injector, with gas fittings for Spectro EOP ICP-OES (75160526).Suitable for routine aqueous analysis, but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7014 Supplier Glass Expansion Supplier No. 308081212
Quartz Torch, single-piece, 2.5 mm injector, with gas fittings for Spectro EOP ICP-OES (75160526).Suitable for routine aqueous analysis, but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001