Learn More
Glass Expansion Quartz Torch with 1.0mm Injector for Agilent 7700/7800/7900/8800/8900
SDP
Catalog No. 501906995
Encompass_Preferred
Quartz Torch, single-piece, 1.0 mm id tapered injector (G3280-80081), for volatile organic solvent analysis with Agilent 7700, 7800, 7850, 7900, 8800, 8900 ICP-MS. Suitable for organic applications with volatile solvents but not inert to samples containing hydrofluoric acid (HF).For volatile organic solvents, also see catalog number 50-190-6994.For samples containing HF see catalog number 30-808-4185.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-6995 Supplier Glass Expansion Supplier No. 308070558
Quartz Torch, single-piece, 1.0 mm id tapered injector (G3280-80081), for volatile organic solvent analysis with Agilent 7700, 7800, 7850, 7900, 8800, 8900 ICP-MS. Suitable for organic applications with volatile solvents but not inert to samples containing hydrofluoric acid (HF).For volatile organic solvents, also see catalog number 50-190-6994.For samples containing HF see catalog number 30-808-4185.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001