Learn More
Glass Expansion Quartz Torch, High TDS, 2.0mm, Shimadzu 7500/8100/9000/9800
SDP
Catalog No. 501906998
Encompass_Preferred
Quartz Torch (High TDS), single-piece, 2.0 mm id injector, for Shimadzu 7500/8100/9000/9800 Series Radial/Axial ICP-OES (211-90942). Suitable for routine aqueous applications but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-6998 Supplier Glass Expansion Supplier No. 308070564
Quartz Torch (High TDS), single-piece, 2.0 mm id injector, for Shimadzu 7500/8100/9000/9800 Series Radial/Axial ICP-OES (211-90942). Suitable for routine aqueous applications but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001