Learn More
Glass Expansion Quartz Torch with 2.0mm Injector for Agilent 7700/7800/7900/8800/8900
SDP
Catalog No. 501907001
Encompass_Preferred
Quartz Torch, single-piece, 2.0 mm id injector, for routine aqueous analysis with Agilent 7700, 7800, 7850, 7900, 8800, 8900 ICP-MS. Suitable for routine aqueous applications but not inert to samples containing hydrofluoric acid (HF).For volatile organic solvents, see catalog number 50-190-6995 or 50-190-6994.For samples containing HF see catalog number 30-808-4185.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7001 Supplier Glass Expansion Supplier No. 308073608
Quartz Torch, single-piece, 2.0 mm id injector, for routine aqueous analysis with Agilent 7700, 7800, 7850, 7900, 8800, 8900 ICP-MS. Suitable for routine aqueous applications but not inert to samples containing hydrofluoric acid (HF).For volatile organic solvents, see catalog number 50-190-6995 or 50-190-6994.For samples containing HF see catalog number 30-808-4185.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001