Learn More
Glass Expansion Quartz Torch Long-life with 90deg bend and 2.3mm Injector for 700-ES or Vista Axial
SDP
Catalog No. 501907004
Encompass_Preferred
Quartz Torch, single-piece, with 90 Deg. bend and 2.3 mm ID injector, for Agilent (Varian) 700-ES or Vista Axial Series ICP-OES. Recommended for routine aqueous analysis but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7004 Supplier Glass Expansion Supplier No. 308080478LL
Quartz Torch, single-piece, with 90 Deg. bend and 2.3 mm ID injector, for Agilent (Varian) 700-ES or Vista Axial Series ICP-OES. Recommended for routine aqueous analysis but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001