Learn More
Glass Expansion Quartz Torch with 1.5mm Injector for Agilent7500
SDP
Catalog No. 501907013
Encompass_Preferred
Quartz Torch, single-piece, 1.5 mm id injector, for aqueous and less-volatile organic solvent analysis with Agilent 7500 ICP-MS. Standard quartz torch suitable for routine aqueous and less-volatile organic solvent applications but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7013 Supplier Glass Expansion Supplier No. 308081211
Quartz Torch, single-piece, 1.5 mm id injector, for aqueous and less-volatile organic solvent analysis with Agilent 7500 ICP-MS. Standard quartz torch suitable for routine aqueous and less-volatile organic solvent applications but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001