Learn More
Glass Expansion Quartz Torch (High Solids) RA/FS13 & 2.3mm Injector for 700-ES or Vista Axial
SDP
Catalog No. 501907016
Encompass_Preferred
Quartz Torch, single-piece (High Solids), 2.3 mm ID injector, for high total dissolved solids (TDS) analysis with Agilent (Varian) 700-ES or Vista Axial ICP-OES. Suitable for routine analysis of high TDS up to 25% but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7016 Supplier Glass Expansion Supplier No. 308081255
Quartz Torch, single-piece (High Solids), 2.3 mm ID injector, for high total dissolved solids (TDS) analysis with Agilent (Varian) 700-ES or Vista Axial ICP-OES. Suitable for routine analysis of high TDS up to 25% but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001