Learn More
Glass Expansion Quartz Torch with 3 slots for Optima 2000/4000/5000/7000 DV
SDP
Catalog No. 501907019
Encompass_Preferred
Quartz Torch, 3-Slot demountable quartz torch standard for PerkinElmer Optima 2000/4000/5000/7000 DV Series ICP-OES (N0772005).Recommended for organic solvent applications, but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7019 Supplier Glass Expansion Supplier No. 308081470
Quartz Torch, 3-Slot demountable quartz torch standard for PerkinElmer Optima 2000/4000/5000/7000 DV Series ICP-OES (N0772005).Recommended for organic solvent applications, but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001