Learn More
Glass Expansion Quartz Torch Elan/NexION
SDP
Catalog No. 501907043
Encompass_Preferred
Quartz Torch, 0-Slot demountable quartz torch standard for PerkinElmer Elan 5000/6x00/9000/DRCs & NexION 300/350 Series ICP-MS (N8012006).Suitable for routine aqueous analysis and is recommended for organic solvent applications, but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7043 Supplier Glass Expansion Supplier No. 308088107
Quartz Torch, 0-Slot demountable quartz torch standard for PerkinElmer Elan 5000/6x00/9000/DRCs & NexION 300/350 Series ICP-MS (N8012006).Suitable for routine aqueous analysis and is recommended for organic solvent applications, but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001