Learn More
Glass Expansion Quartz Torch with 3 slots for Optima 8x00
SDP
Catalog No. 501907195
Encompass_Preferred
Quartz Torch, 3-Slot demountable quartz torch standard for PerkinElmer Optima 8x00 Series ICP-OES (N0780132).Recommended for organic solvent applications, but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7195 Supplier Glass Expansion Supplier No. 318083256
Quartz Torch, 3-Slot demountable quartz torch standard for PerkinElmer Optima 8x00 Series ICP-OES (N0780132).Recommended for organic solvent applications, but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001