Learn More
Glass Expansion Quartz Torch for SpectroBlue TI
SDP
Catalog No. 501907225
Encompass_Preferred
Quartz Torch, semi-demountable torch for Spectro (Ametek) SpectroBlue TI ICP-OES (48205054).Suitable for routine aqueous and organic applications, but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7225 Supplier Glass Expansion Supplier No. 318083468
Quartz Torch, semi-demountable torch for Spectro (Ametek) SpectroBlue TI ICP-OES (48205054).Suitable for routine aqueous and organic applications, but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001