Learn More
Glass Expansion Quartz Torch with single slot for Optima 8x00 (N0780130, N0780131, 8003-0344)
SDP
Catalog No. 501907194
Encompass_Preferred
Quartz Torch, 1-Slot demountable quartz torch standard for PerkinElmer Optima 8x00 Series ICP-OES (N0780130).Standard torch for routine aqueous applications, but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7194 Supplier Glass Expansion Supplier No. 318083255
Quartz Torch, 1-Slot demountable quartz torch standard for PerkinElmer Optima 8x00 Series ICP-OES (N0780130).Standard torch for routine aqueous applications, but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001