Learn More
Glass Expansion Quartz Torch for Spectro Blue SOP
SDP
Catalog No. 501907218
Encompass_Preferred
Quartz Torch, semi-demountable torch for Spectro (Ametek) SpectroBlue SOP ICP-OES (48205045).Suitable for routine aqueous and organic applications, but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7218 Supplier Glass Expansion Supplier No. 318083405
Quartz Torch, semi-demountable torch for Spectro (Ametek) SpectroBlue SOP ICP-OES (48205045).Suitable for routine aqueous and organic applications, but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001